Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Thiophenesulfonamide 98.0+%, TCI America™
SDP

Catalog No. T28515G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T28515G 5 g
T28511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T28515G Supplier TCI America Supplier No. T28515G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6339-87-3
Molecular Formula C4H5NO2S2
Molecular Weight (g/mol) 163.209
MDL Number MFCD00185853
InChI Key KTFDYVNEGTXQCV-UHFFFAOYSA-N
PubChem CID 72881
ChEBI CHEBI:79403
IUPAC Name thiophene-2-sulfonamide
SMILES C1=CSC(=C1)S(=O)(=O)N

Specifications

Melting Point 146°C
Quantity 5 g
Formula Weight 163.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Thiophenesulfonamide
RTECSNumber XN0294000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.