CAS RN 634207-53-7
IUPAC Name:
6-{6-amino-8-[(4-chlorophenyl)sulfanyl]-9H-purin-9-yl}-2-hydroxy-7-methoxy-hexahydro-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinin-2-one sodium
Synonyms:
6-{6-amino-8-[(4-chlorophenyl)sulfanyl]purin-9-yl}-2-hydroxy-7-methoxy-tetrahydro-4H-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinin-2-one sodium
Molecular Weight (g/mol):
508.83
Molecular Formula:
C17H17ClN5NaO6PS
InChi Key:
IORFFVUNBNOFAO-UHFFFAOYNA-N
SMILES:
[Na].COC1C(OC2COP(O)(=O)OC12)N1C(SC2=CC=C(Cl)C=C2)=NC2=C(N)N=CN=C12
1
–
1
of
1
results
1
R&D Systems™ 8-CPT-2Me-cAMP, sodium salt
CAS: 634207-53-7 Molecular Formula: C17H17ClN5NaO6PS