accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaNaHONHNOSOOOSOOOSOOO(Z)

CAS RN 63451-35-4

IUPAC Name: trisodium 3-hydroxy-4-{2-[(1Z)-2-oxo-4-sulfonato-1,2-dihydronaphthalen-1-ylidene]hydrazin-1-yl}naphthalene-2,7-disulfonate
Synonyms: hydroxynaphthol blue
Molecular Weight (g/mol): 620.46
Molecular Formula: C20H11N2Na3O11S3
InChi Key: JQECMNGWLIFQLL-QRQZZYIYSA-K
SMILES: [Na+].[Na+].[Na+].OC1=C(C=C2C=C(C=CC2=C1N\N=C1/C(=O)C=C(C2=CC=CC=C12)S([O-])(=O)=O)S([O-])(=O)=O)S([O-])(=O)=O
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Color 
  • (1)
  • (3)
Grade 
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (1)
pH 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)