Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxynaphthol Blue, Indicator for Metal Titration, Solstice

Catalog No. 60046314 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-314 50 g Glass Bottle
60-046-313 10 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-046-314 Supplier Solstice Supplier No. 3393650G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Indicator for metal titration

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63451-35-4
Molecular Formula C20H11N2Na3O11S3
Molecular Weight (g/mol) 620.46
MDL Number MFCD00036393
InChI Key JQECMNGWLIFQLL-UHFFFAOYSA-K
PubChem CID 44134769
IUPAC Name trisodium;3-hydroxy-4-[2-(2-oxo-4-sulfonatonaphthalen-1-ylidene)hydrazinyl]naphthalene-2,7-disulfonate
SMILES C1=CC=C2C(=C1)C(=CC(=O)C2=NNC3=C4C=CC(=CC4=CC(=C3O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+]

Specifications

Infrared Spectrum Conforms to structure
Packaging Glass Bottle
UN Number NONH for all modes of transport
Quantity 50 g
Physical Form Liquid
Formula Weight 620.47g/mol
Chemical Name or Material Hydroxynaphthol blue
RTECSNumber 264-197-7
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.