accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNNNH2NOH3C

CAS RN 634924-89-3

IUPAC Name: 8-ethoxy-9-ethyl-9H-purin-6-amine
Synonyms: 8-ethoxy-9-ethylpurin-6-amine
Molecular Weight (g/mol): 207.24
Molecular Formula: C9H13N5O
InChi Key: QUGDTMONBLMLLD-UHFFFAOYSA-N
SMILES: CCOC1=NC2=C(N)N=CN=C2N1CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)