accessibility menu, dialog, popup

UserName

Viewing profile as user:
OBrClF

CAS RN 63529-30-6

IUPAC Name: 2-bromo-1-(3-chloro-4-fluorophenyl)ethan-1-one
Synonyms: 2-bromo-1-(3-chloro-4-fluorophenyl)ethanone
Molecular Weight (g/mol): 251.48
Molecular Formula: C8H5BrClFO
InChi Key: JOCPGHGWUUBURW-UHFFFAOYSA-N
SMILES: FC1=C(Cl)C=C(C=C1)C(=O)CBr
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)