Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-4-fluorophenacyl Bromide 98.0+%, TCI America™
SDP

Catalog No. C27525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27525G 5 g
C27521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C27525G Supplier TCI America Supplier No. C27525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 63529-30-6
Molecular Formula C8H5BrClFO
Molecular Weight (g/mol) 251.48
MDL Number MFCD05154991
InChI Key JOCPGHGWUUBURW-UHFFFAOYSA-N
Synonym 2-bromo-1-3-chloro-4-fluorophenyl ethanone, 2-bromo-3'-chloro-4'-fluoroacetophenone, 3-chloro-4-fluorophenacyl bromide, 2-bromo-1-3-chloro-4-fluorophenyl ethan-1-one, ethanone, 2-bromo-1-3-chloro-4-fluorophenyl, acmc-209ng8, 3-chloro-4-fluoro-phenacyl bromide, 2-bromo-3-chloro-4-fluoroacetophenone
PubChem CID 2782785
IUPAC Name 2-bromo-1-(3-chloro-4-fluorophenyl)ethan-1-one
SMILES FC1=C(Cl)C=C(C=C1)C(=O)CBr

Specifications

Melting Point 36°C
Quantity 5 g
UN Number 3261
Formula Weight 251.48
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-4-fluorophenacyl Bromide
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.