Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-3'-chloro-4'-fluoroacetophenone, 96%, Thermo Scientific™

Catalog No. AAH6185203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6185203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6185203 Supplier Thermo Scientific Chemicals Supplier No. H6185203

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63529-30-6
Molecular Formula C8H5BrClFO
Molecular Weight (g/mol) 251.48
MDL Number MFCD05154991
InChI Key JOCPGHGWUUBURW-UHFFFAOYSA-N
Synonym 2-bromo-1-3-chloro-4-fluorophenyl ethanone, 2-bromo-3'-chloro-4'-fluoroacetophenone, 3-chloro-4-fluorophenacyl bromide, 2-bromo-1-3-chloro-4-fluorophenyl ethan-1-one, ethanone, 2-bromo-1-3-chloro-4-fluorophenyl, acmc-209ng8, 3-chloro-4-fluoro-phenacyl bromide, 2-bromo-3-chloro-4-fluoroacetophenone
PubChem CID 2782785
IUPAC Name 2-bromo-1-(3-chloro-4-fluorophenyl)ethanone
SMILES FC1=C(Cl)C=C(C=C1)C(=O)CBr

Specifications

Melting Point 33°C to 36°C
Flash Point >110°C (230°F)
Quantity 1 g
Formula Weight 251.48
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 2-Bromo-3'-chloro-4'-fluoroacetophenone
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.