accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOSOOCl

CAS RN 63545-54-0

IUPAC Name: 3-[4-(chlorosulfonyl)phenyl]propanoic acid
Synonyms: 3-[4-(chlorosulfonyl)phenyl]propanoic acid
Molecular Weight (g/mol): 248.68
Molecular Formula: C9H9ClO4S
InChi Key: INJMPXHPNFJMLG-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=CC=C(C=C1)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results