accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaCH3ONNCH3HNNOSOOOSOOOCH3(E/Z)(Z)

CAS RN 6358-29-8

IUPAC Name: disodium (8Z)-8-(2-{4'-[2-(4-ethoxyphenyl)diazen-1-yl]-3,3'-dimethyl-[1,1'-biphenyl]-4-yl}hydrazin-1-ylidene)-7-oxo-7,8-dihydronaphthalene-1,3-disulfonate
Synonyms: disodium (8Z)-8-(2-{4'-[2-(4-ethoxyphenyl)diazen-1-yl]-3,3'-dimethyl-[1,1'-biphenyl]-4-yl}hydrazin-1-ylidene)-7-oxonaphthalene-1,3-disulfonate
Molecular Weight (g/mol): 704.68
Molecular Formula: C32H26N4Na2O8S2
InChi Key: ZOYGJDNPTASPDH-DQQIWZDQSA-L
SMILES: [Na+].[Na+].CCOC1=CC=C(C=C1)N=NC1=CC=C(C=C1C)C1=CC=C(N\N=C2/C(=O)C=CC3=CC(=CC(=C23)S([O-])(=O)=O)S([O-])(=O)=O)C(C)=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results