Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Direct Fast Red 3B, TCI America™
SDP

Catalog No. D147225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D147225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D147225G Supplier TCI America Supplier No. D147225G

Chemical Identifiers

CAS 6358-29-8
Molecular Formula C32H28N4Na2O8S2
Molecular Weight (g/mol) 706.696
MDL Number MFCD00059129
InChI Key DWOCRJJONVWKDP-DQQIWZDQSA-N
Synonym Direct Scarlet 3B, Direct Red 39
PubChem CID 131882482
IUPAC Name (8Z)-8-[[4-[4-[(4-ethoxyphenyl)diazenyl]-3-methylphenyl]-2-methylphenyl]hydrazinylidene]-7-oxonaphthalene-1,3-disulfonic acid;sodium
SMILES CCOC1=CC=C(C=C1)N=NC2=C(C=C(C=C2)C3=CC(=C(C=C3)NN=C4C(=O)C=CC5=CC(=CC(=C54)S(=O)(=O)O)S(=O)(=O)O)C)C.[Na].[Na]

Specifications

Color Purple-Red
Quantity 25 g
Formula Weight 704.68
Physical Form Crystalline Powder
Chemical Name or Material Direct Fast Red 3B
RTECSNumber QJ6476000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.