CAS RN 63598-32-3
IUPAC Name:
N-{6-methoxy-2-phenyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-8-ylidene}hydroxylamine
Synonyms:
N-{6-methoxy-2-phenyl-hexahydropyrano[3,2-d][1,3]dioxin-8-ylidene}hydroxylamine
Molecular Weight (g/mol):
279.29
Molecular Formula:
C14H17NO5
InChi Key:
ZVPSUEFQWUNIMY-UHFFFAOYNA-N
SMILES:
COC1CC(=NO)C2OC(OCC2O1)C1=CC=CC=C1
1
–
1
of
1
results
1
Methyl 4,6-O-Benzylidene-2-deoxy-alpha-D-erythro-hexopyranosid-3-ulose Oxime 98.0+%, TCI America™
CAS: 63598-32-3 Molecular Formula: C14H17NO5 Molecular Weight (g/mol): 279.29 MDL Number: MFCD15072181 InChI Key: ZVPSUEFQWUNIMY-UHFFFAOYNA-N PubChem CID: 129441492 IUPAC Name: N-{6-methoxy-2-phenyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-8-ylidene}hydroxylamine SMILES: COC1CC(=NO)C2OC(OCC2O1)C1=CC=CC=C1