Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4,6-O-Benzylidene-2-deoxy-alpha-D-erythro-hexopyranosid-3-ulose Oxime 98.0+%, TCI America™
SDP

Catalog No. M20811G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M20811G Supplier TCI America Supplier No. M20811G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 63598-32-3
Molecular Formula C14H17NO5
Molecular Weight (g/mol) 279.29
MDL Number MFCD15072181
InChI Key ZVPSUEFQWUNIMY-UHFFFAOYNA-N
PubChem CID 129441492
IUPAC Name N-{6-methoxy-2-phenyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-8-ylidene}hydroxylamine
SMILES COC1CC(=NO)C2OC(OCC2O1)C1=CC=CC=C1

Specifications

Melting Point 200°C
Color White
Quantity 1 g
Formula Weight 279.29
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4,6-O-Benzylidene-2-deoxy-alpha-D-erythro-hexopyranosid-3-ulose Oxime
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.