accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHSHNO

CAS RN 636-26-0

IUPAC Name: 5-methyl-2-sulfanylidene-1,2,3,4-tetrahydropyrimidin-4-one
Synonyms: thiothymin
Molecular Weight (g/mol): 142.18
Molecular Formula: C5H6N2OS
InChi Key: ZLAQATDNGLKIEV-UHFFFAOYSA-N
SMILES: CC1=CNC(=S)NC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results