accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2(S)HNOOOOHCl

CAS RN 6366-70-7

IUPAC Name: benzyl (2S)-2-amino-6-{[(2-phenylacetyl)oxy]amino}hexanoate hydrochloride
Synonyms: benzyl (2S)-2-amino-6-{[(2-phenylacetyl)oxy]amino}hexanoate hydrochloride|H-LYS(Z)-OBZL HCL
Molecular Weight (g/mol): 406.91
Molecular Formula: C21H27ClN2O4
InChi Key: NWSFLBWTZZJOAF-FYZYNONXSA-N
SMILES: Cl.N[C@@H](CCCCNOC(=O)CC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)