accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaOONHNOHNSOOO(E)

CAS RN 6369-32-0

IUPAC Name: disodium 2-{2-[(2E)-1-oxo-7-(phenylamino)-3-sulfonato-1,2-dihydronaphthalen-2-ylidene]hydrazin-1-yl}benzoate
Synonyms: disodium 2-{2-[(2E)-1-oxo-7-(phenylamino)-3-sulfonatonaphthalen-2-ylidene]hydrazin-1-yl}benzoate
Molecular Weight (g/mol): 507.43
Molecular Formula: C23H15N3Na2O6S
InChi Key: GDVUVRLDXFWCSM-CXJWIYSGSA-L
SMILES: [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1N\N=C1/C(=O)C2=CC(NC3=CC=CC=C3)=CC=C2C=C1S([O-])(=O)=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results