accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHSOOClClCl

CAS RN 6378-25-2

IUPAC Name: 2,4,5-trichlorobenzene-1-sulfonic acid
Synonyms: 2,4,5-trichlorobenzenesulfonic acid
Molecular Weight (g/mol): 261.5
Molecular Formula: C6H3Cl3O3S
InChi Key: LEDKKDPOPIKMSZ-UHFFFAOYSA-N
SMILES: OS(=O)(=O)C1=CC(Cl)=C(Cl)C=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results