CAS RN 6378-65-0
IUPAC Name:
hexyl hexanoate
Synonyms:
hexyl hexanoate
Molecular Weight (g/mol):
200.32
Molecular Formula:
C12H24O2
InChi Key:
NCDCLPBOMHPFCV-UHFFFAOYSA-N
SMILES:
CCCCCCOC(=O)CCCCC
1
–
2
of
2
results
1
Hexyl Hexanoate 98.0+%, TCI America™
CAS: 6378-65-0 Molecular Formula: C12H24O2 Molecular Weight (g/mol): 200.32 MDL Number: MFCD00053808 InChI Key: NCDCLPBOMHPFCV-UHFFFAOYSA-N Synonym: Hexanoic Acid Hexyl Ester PubChem CID: 22873 IUPAC Name: hexyl hexanoate SMILES: CCCCCCOC(=O)CCCCC