Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexyl Hexanoate 98.0+%, TCI America™
SDP

Catalog No. C1514500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1514500ML 500 mL
C151425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1514500ML Supplier TCI America Supplier No. C1514500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6378-65-0
Molecular Formula C12H24O2
Molecular Weight (g/mol) 200.32
MDL Number MFCD00053808
InChI Key NCDCLPBOMHPFCV-UHFFFAOYSA-N
Synonym Hexanoic Acid Hexyl Ester
PubChem CID 22873
IUPAC Name hexyl hexanoate
SMILES CCCCCCOC(=O)CCCCC

Specifications

Color Colorless
Boiling Point 244°C
Quantity 500 mL
Formula Weight 200.32
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexyl Hexanoate
EINECSNumber (2)-0760
RTECSNumber MO8385000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.