accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6389-79-3

IUPAC Name: methyl methyl(phenyl)phosphinate
Synonyms: methyl methyl(phenyl)phosphinate
Molecular Weight (g/mol): 170.15
Molecular Formula: C8H11O2P
InChi Key: RTAAKNLNWIHGGX-UHFFFAOYNA-N
SMILES: COP(C)(=O)C1=CC=CC=C1
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)