accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 63927-23-1

IUPAC Name: 5-bromo-8-nitroisoquinoline
Synonyms: 5-bromo-8-nitroisoquinoline
Molecular Weight (g/mol): 253.06
Molecular Formula: C9H5BrN2O2
InChi Key: ULGOLOXWHJEZNZ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C2C=NC=CC2=C(Br)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results