accessibility menu, dialog, popup

UserName

Viewing profile as user:
PHI

CAS RN 6396-08-3

IUPAC Name: triphenylphosphane hydroiodide
Synonyms: triphenylphosphine hydroiodide
Molecular Weight (g/mol): 390.2
Molecular Formula: C18H16IP
InChi Key: POXSDSRWVJZWCN-UHFFFAOYSA-N
SMILES: I.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results