CAS RN 64-18-6
Formic Acid, 99.0+%, Optima™ LC/MS Grade, Fisher Chemical™
CAS: 64-18-6 Molecular Formula: CH2O2 MDL Number: 3297 InChI Key: BDAGIHXWWSANSR-UHFFFAOYSA-N Synonym: methanoic acid,formylic acid,aminic acid,bilorin,hydrogen carboxylic acid,formisoton,myrmicyl,formira,acide formique,collo-bueglatt PubChem CID: 284 ChEBI: CHEBI:30751 IUPAC Name: formic acid
Formic Acid ACS, 98%, MilliporeSigma™
CAS: 64-18-6 Molecular Formula: CH2O2 Molecular Weight (g/mol): 46.025 InChI Key: BDAGIHXWWSANSR-UHFFFAOYSA-N Synonym: methanoic acid,formylic acid,aminic acid,bilorin,hydrogen carboxylic acid,formisoton,myrmicyl,formira,acide formique,collo-bueglatt PubChem CID: 284 ChEBI: CHEBI:30751 IUPAC Name: formic acid SMILES: C(=O)O
Formic Acid, 88% (Certified ACS), Fisher Chemical™
CAS: 64-18-6 Molecular Formula: CH2O2 Molecular Weight (g/mol): 46.025 MDL Number: MFCD00003297 InChI Key: BDAGIHXWWSANSR-UHFFFAOYSA-N Synonym: methanoic acid,formylic acid,aminic acid,bilorin,hydrogen carboxylic acid,formisoton,myrmicyl,formira,acide formique,collo-bueglatt PubChem CID: 284 ChEBI: CHEBI:30751 IUPAC Name: formic acid SMILES: C(=O)O
Formic Acid 98.0+%, TCI America™
CAS: 64-18-6 Molecular Formula: CH2O2 Molecular Weight (g/mol): 46.025 MDL Number: MFCD00003297 InChI Key: BDAGIHXWWSANSR-UHFFFAOYSA-N Synonym: methanoic acid,formylic acid,aminic acid,bilorin,hydrogen carboxylic acid,formisoton,myrmicyl,formira,acide formique,collo-bueglatt PubChem CID: 284 ChEBI: CHEBI:30751 IUPAC Name: formic acid SMILES: C(=O)O
Formic Acid, 88% (Laboratory), Fisher Chemical™
CAS: 64-18-6 Molecular Formula: CH2O2 Molecular Weight (g/mol): 46.025 MDL Number: MFCD00003297 InChI Key: BDAGIHXWWSANSR-UHFFFAOYSA-N Synonym: methanoic acid,formylic acid,aminic acid,bilorin,hydrogen carboxylic acid,formisoton,myrmicyl,formira,acide formique,collo-bueglatt PubChem CID: 284 ChEBI: CHEBI:30751 IUPAC Name: formic acid SMILES: C(=O)O
Formic acid, 99%, for analysis
CAS: 64-18-6 Molecular Formula: CH2O2 Molecular Weight (g/mol): 46.025 MDL Number: MFCD00003297 InChI Key: BDAGIHXWWSANSR-UHFFFAOYSA-N Synonym: methanoic acid,formylic acid,aminic acid,bilorin,hydrogen carboxylic acid,formisoton,myrmicyl,formira,acide formique,collo-bueglatt PubChem CID: 284 ChEBI: CHEBI:30751 IUPAC Name: formic acid SMILES: C(=O)O
Formic acid, 98+%, pure
CAS: 64-18-6 Molecular Formula: CH2O2 Molecular Weight (g/mol): 46.025 InChI Key: BDAGIHXWWSANSR-UHFFFAOYSA-N Synonym: methanoic acid,formylic acid,aminic acid,bilorin,hydrogen carboxylic acid,formisoton,myrmicyl,formira,acide formique,collo-bueglatt PubChem CID: 284 ChEBI: CHEBI:30751 IUPAC Name: formic acid SMILES: C(=O)O
Formic acid, 97%
CAS: 64-18-6 Molecular Formula: CH2O2 Molecular Weight (g/mol): 46.025 MDL Number: MFCD00003297 InChI Key: BDAGIHXWWSANSR-UHFFFAOYSA-N Synonym: methanoic acid,formylic acid,aminic acid,bilorin,hydrogen carboxylic acid,formisoton,myrmicyl,formira,acide formique,collo-bueglatt PubChem CID: 284 ChEBI: CHEBI:30751 IUPAC Name: formic acid SMILES: C(=O)O
Formic acid, ACS, 96+%
CAS: 64-18-6 Molecular Formula: CH2O2 Molecular Weight (g/mol): 46.025 MDL Number: MFCD00003297 InChI Key: BDAGIHXWWSANSR-UHFFFAOYSA-N Synonym: methanoic acid,formylic acid,aminic acid,bilorin,hydrogen carboxylic acid,formisoton,myrmicyl,formira,acide formique,collo-bueglatt PubChem CID: 284 ChEBI: CHEBI:30751 IUPAC Name: formic acid SMILES: C(=O)O
Formic Acid, 98% (ACS), GR, MilliporeSigma™
CAS: 64-18-6 Molecular Formula: CH2O2 Molecular Weight (g/mol): 46.025 InChI Key: BDAGIHXWWSANSR-UHFFFAOYSA-N Synonym: methanoic acid,formylic acid,aminic acid,bilorin,hydrogen carboxylic acid,formisoton,myrmicyl,formira,acide formique,collo-bueglatt PubChem CID: 284 ChEBI: CHEBI:30751 IUPAC Name: formic acid SMILES: C(=O)O
Formic acid, ACS, 88+%
CAS: 64-18-6 Molecular Formula: CH2O2 Molecular Weight (g/mol): 46.025 MDL Number: MFCD00003297 InChI Key: BDAGIHXWWSANSR-UHFFFAOYSA-N Synonym: methanoic acid,formylic acid,aminic acid,bilorin,hydrogen carboxylic acid,formisoton,myrmicyl,formira,acide formique,collo-bueglatt PubChem CID: 284 ChEBI: CHEBI:30751 IUPAC Name: formic acid SMILES: C(=O)O
Formic acid, 88%, ACS reagent
CAS: 64-18-6 Molecular Formula: CH2O2 Molecular Weight (g/mol): 46.025 InChI Key: BDAGIHXWWSANSR-UHFFFAOYSA-N Synonym: methanoic acid,formylic acid,aminic acid,bilorin,hydrogen carboxylic acid,formisoton,myrmicyl,formira,acide formique,collo-bueglatt PubChem CID: 284 ChEBI: CHEBI:30751 IUPAC Name: formic acid SMILES: C(=O)O
Formic Acid, 96%, Reagent, ACS, 96%, Spectrum™ Chemical
CAS: 64-18-6 Molecular Formula: CH2O2 Molecular Weight (g/mol): 46.03 InChI Key: BDAGIHXWWSANSR-UHFFFAOYSA-N IUPAC Name: formic acid SMILES: OC=O
Formic Acid, FCC, 85%, Spectrum™ Chemical
CAS: 64-18-6 Molecular Formula: CH2O2 Molecular Weight (g/mol): 46.03 InChI Key: BDAGIHXWWSANSR-UHFFFAOYSA-N IUPAC Name: formic acid SMILES: OC=O