Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formic acid, ACS, 96+%

Catalog No. AA36617A3
Change view
Click to view available options
Quantity:
500 g
2 kg
4 x 500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA36617A3 2 kg
AA3661736 500 g
AA3661757 4 x 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA36617A3 Supplier Thermo Scientific Chemicals Supplier No. 036617A3
Add to Cart

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 64-18-6 | CH2O2 | 46.025 g/mol

Used as a hydrogen source, as a reducing agent and is used in tanning and dye fixing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-18-6
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.025
MDL Number MFCD00003297
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
PubChem CID 284
ChEBI CHEBI:30751
IUPAC Name formic acid
SMILES C(=O)O

Specifications

Melting Point 7°C to 9°C
Boiling Point 100°C to 101°C
Color Colorless
Density 1.220 g/mL
Quantity 2 kg
Flash Point 50°C (156°F)
Physical Form Liquid
Refractive Index 1.371
Assay Percent Range >96+%
Sensitivity Hygroscopic
Linear Formula HCOOH
UN Number UN1779
Beilstein 1209246
Merck Index 14,4241
Solubility Information Miscible with water. Partially miscible in toluene,xylene and benzene.
Formula Weight 46.03
Percent Purity ≥96%
Grade ACS
Chemical Name or Material Formic acid, For analysis ACS
Show More Show Less
Hazard Category H226-H302-H314-H331
Hazard Statement GHS H Statement
H331-H314-H318-H226-H302
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P311-P312-P330-P331-P363-P372-P374-P380-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: FORMIC ACID
EINECSNumber 200-579-1
RTECSNumber LQ4900000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.