Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formic Acid ACS, 98%, MilliporeSigma™
GSA_VA

Catalog No. MFX04405 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mL
1 L
25 L
Packaging:
Metal Drum w/Bag
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MFX04405 500 mL Poly Bottle
MFX04407 1 L Poly Bottle
MFX044021 25 L Metal Drum w/Bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MFX04405 Supplier MilliporeSigma Supplier No. US1FX0440/5
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-18-6
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.025
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
PubChem CID 284
ChEBI CHEBI:30751
IUPAC Name formic acid
SMILES C(=O)O

Specifications

Quantity 500 mL
Flash Point 50°C(122°F)
Packaging Poly Bottle
Physical Form Liquid
Assay Percent Range 98%
Linear Formula HCOOH
Formula Weight 46.3
Grade ACS/NF/USP
Chemical Name or Material Formic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.