accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ClH2NCl

CAS RN 64063-37-2

IUPAC Name: 2,6-dichloro-3-methylaniline
Synonyms: 2,6-dichloro-3-methylaniline
Molecular Weight (g/mol): 176.04
Molecular Formula: C7H7Cl2N
InChi Key: AYVZXDFCFQSWJD-UHFFFAOYSA-N
SMILES: CC1=CC=C(Cl)C(N)=C1Cl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results