accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 641-74-7

IUPAC Name: hexahydrofuro[3,2-b]furan-3,6-diol
Synonyms: isosorbide
Molecular Weight (g/mol): 146.14
Molecular Formula: C6H10O4
InChi Key: KLDXJTOLSGUMSJ-UHFFFAOYNA-N
SMILES: OC1COC2C(O)COC12
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)