accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6410-10-2

IUPAC Name: (1Z)-1-[2-(4-nitrophenyl)hydrazin-1-ylidene]-1,2-dihydronaphthalen-2-one
Synonyms: para red
Molecular Weight (g/mol): 293.28
Molecular Formula: C16H11N3O3
InChi Key: ACHQDMGGRZPFAW-VLGSPTGOSA-N
SMILES: [O-][N+](=O)C1=CC=C(N\N=C2/C(=O)C=CC3=CC=CC=C23)C=C1
Quantity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results