Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dihydro-5-methyl-2-(4-nitrophenyl)-3H-pyrazol-3-one 95%, Thermo Scientific™

Catalog No. AC407620250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC407620250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC407620250 Supplier Thermo Scientific Chemicals Supplier No. 407620250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6410-10-2
Molecular Formula C16H11N3O3
Molecular Weight (g/mol) 293.28
InChI Key ACHQDMGGRZPFAW-VLGSPTGOSA-N
Synonym pigment red 1, federal red, para toner, label red, eljon para red, para red light, para red toner, para toner red, pigment red zh, lutetia red b
PubChem CID 6076344
IUPAC Name (1E)-1-[(4-nitrophenyl)hydrazinylidene]naphthalen-2-one
SMILES C1=CC=C2C(=C1)C=CC(=O)C2=NNC3=CC=C(C=C3)[N+](=O)[O-]

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 220°C to 225°C
Color Yellow to Brown
Infrared Spectrum Authentic
Quantity 25 g
Formula Weight 219.2
Percent Purity ≥94%
Physical Form Crystalline Powder
Chemical Name or Material 2, 4-Dihydro-5-methyl-2-(4-nitrophenyl)-3H-pyrazol-3-one, 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.