Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Para Red, TCI America™
SDP

Catalog No. P058625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P058625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P058625G Supplier TCI America Supplier No. P058625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6410-10-2
Molecular Formula C16H11N3O3
Molecular Weight (g/mol) 293.28
MDL Number MFCD00003908
InChI Key ACHQDMGGRZPFAW-VLGSPTGOSA-N
Synonym 1-(4-Nitrophenylazo)-2-naphthol
PubChem CID 6076344
IUPAC Name (1E)-1-[(4-nitrophenyl)hydrazinylidene]naphthalen-2-one
SMILES C1=CC=C2C(=C1)C=CC(=O)C2=NNC3=CC=C(C=C3)[N+](=O)[O-]

Specifications

Melting Point 252°C
Quantity 25 g
Physical Form Crystal/Powder
Formula Weight 293.28
Chemical Name or Material Para Red
EINECSNumber (5)-3207
RTECSNumber QL4510000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.