accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CPH3CPCH3CH3

CAS RN 6411-21-8

IUPAC Name: [2-(diethylphosphanyl)ethyl]diethylphosphane
Synonyms: [2-(diethylphosphanyl)ethyl]diethylphosphane
Molecular Weight (g/mol): 206.25
Molecular Formula: C10H24P2
InChi Key: MIOCUERTSIJEDP-UHFFFAOYSA-N
SMILES: CCP(CC)CCP(CC)CC
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)