accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHClONHClCl

CAS RN 642-84-2

IUPAC Name: 5-chloro-N-(3,4-dichlorophenyl)-2-hydroxybenzamide
Synonyms: 5-chloro-N-(3,4-dichlorophenyl)-2-hydroxybenzamide
Molecular Weight (g/mol): 316.56
Molecular Formula: C13H8Cl3NO2
InChi Key: RSJBLPJKXGNMFW-UHFFFAOYSA-N
SMILES: OC1=CC=C(Cl)C=C1C(=O)NC1=CC=C(Cl)C(Cl)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results