accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 64208-32-8

IUPAC Name: 4-[3-(tert-butylamino)-2-hydroxypropoxy]-2,3-dihydro-1H-1,3-benzodiazol-2-one hydrochloride
Synonyms: 4-[3-(tert-butylamino)-2-hydroxypropoxy]-1,3-dihydro-1,3-benzodiazol-2-one hydrochloride
Molecular Weight (g/mol): 315.8
Molecular Formula: C14H22ClN3O3
InChi Key: YQVFCYCTITZLSX-UHFFFAOYNA-N
SMILES: Cl.CC(C)(C)NCC(O)COC1=C2NC(=O)NC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results