Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ CGP 12177 hydrochloride
GREENER_CHOICESDP

Catalog No. 113450 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-345-0 50 mg
11-341-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 11-345-0 Supplier R&D Systems™ Supplier No. 1134/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. β3 partial agonist. β1/β2 antagonist

Partial agonist at β3-adrenoceptors. β1- and β2-adrenoceptor antagonist (Ki values are 0.9, 4 and 88 nM for β1, β2 and β3 receptors respectively).

Chemical Identifiers

CAS 64208-32-8
Molecular Formula C14H22ClN3O3

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 315.8
Purity >98%
Molecular Weight (g/mol) 315.798
Chemical Name or Material CGP 12177 hydrochloride
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.