accessibility menu, dialog, popup

UserName

Viewing profile as user:
C

CAS RN 64365-11-3

IUPAC Name: carbon
Synonyms: carbon carbone carbon atom carbono carbonium kohlenstoff atomic carbon
Molecular Weight (g/mol): 12.01
Molecular Formula: C
InChi Key: OKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES: [C]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)