accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HO

CAS RN 644-35-9

IUPAC Name: 2-propylphenol
Synonyms: O-propylphenol
Molecular Weight (g/mol): 136.19
Molecular Formula: C9H12O
InChi Key: LCHYEKKJCUJAKN-UHFFFAOYSA-N
SMILES: CCCC1=CC=CC=C1O
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results