accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 64415-15-2

IUPAC Name: 1-hydroxy-4-sulfamoylnaphthalene-2-carboxylic acid
Synonyms: 1-hydroxy-4-sulfamoylnaphthalene-2-carboxylic acid
Molecular Weight (g/mol): 267.26
Molecular Formula: C11H9NO5S
InChi Key: OTDVKOVLSPJORY-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=C2C=CC=CC2=C(O)C(=C1)C(O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results