CAS RN 64419-92-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Necrostatin-1, Inactive Control
A cell-permeable N-demethylated thiohydantoin analog of Nec-1 that is devoid of anti-necroptotic properties and serves as a suitable inactive control.