accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OHO

CAS RN 646-07-1

IUPAC Name: 4-methylpentanoic acid
Synonyms: 4-methyl valeric acid isocaproate isohexanoic acid (mixed isomers) isocaproic acid
Molecular Weight (g/mol): 116.16
Molecular Formula: C6H12O2
InChi Key: FGKJLKRYENPLQH-UHFFFAOYSA-N
SMILES: CC(C)CCC(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (9)
Boiling Point 
  • (3)
  • (2)
  • (2)
  • (3)
Percent Purity 
  • (1)
  • (2)
  • (7)
Quantity 
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (1)
  • (3)
  • (5)
  • (2)
  • (9)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (7)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (5)
  • (1)