Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylvaleric Acid (contains 3-Methylvaleric Acid) 75.0+%, TCI America™
SDP

Catalog No. M045725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M045725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M045725ML Supplier TCI America Supplier No. M045725ML

Chemical Identifiers

CAS 646-07-1
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00002803
InChI Key FGKJLKRYENPLQH-UHFFFAOYSA-N
Synonym 4-methylvaleric acid, isocaproic acid, isohexanoic acid, pentanoic acid, 4-methyl, isobutylacetic acid, 4-methyl-pentanoic acid, valeric acid, 4-methyl, 4-methyl valeric acid, isocaproate, 4,4-dimethylbutanoic acid
PubChem CID 12587
ChEBI CHEBI:74903
IUPAC Name 4-methylpentanoic acid
SMILES CC(C)CCC(=O)O

Specifications

Color Yellow
Boiling Point 201°C
Quantity 25 mL
UN Number 2810
Formula Weight 116.16
Percent Purity ≥75.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methylvaleric Acid (contains 3-Methylvaleric Acid)
EINECSNumber (2)-0608
RTECSNumber NR2975000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.