accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 64689-70-9

IUPAC Name: 1-bromo-2-(methoxymethyl)naphthalene
Synonyms: 1-bromo-2-(methoxymethyl)naphthalene
Molecular Weight (g/mol): 251.12
Molecular Formula: C12H11BrO
InChi Key: ORYVAZPUJFACFR-UHFFFAOYSA-N
SMILES: COCC1=CC=C2C=CC=CC2=C1Br
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results