accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 64691-70-9

IUPAC Name: 2-{2-[(2-{[2-(pyridin-2-yl)ethyl]sulfanyl}ethyl)sulfanyl]ethyl}pyridine
Synonyms: 2-{2-[(2-{[2-(pyridin-2-yl)ethyl]sulfanyl}ethyl)sulfanyl]ethyl}pyridine
Molecular Weight (g/mol): 304.47
Molecular Formula: C16H20N2S2
InChi Key: YDPDVMHQFYTZDI-UHFFFAOYSA-N
SMILES: C(CSCCC1=CC=CC=N1)SCCC1=CC=CC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results