CAS RN 64742-49-0
Petroleum ether, extra pure, boiling range 100-140°C
CAS: 64742-49-0
Petroleum ether, for analysis, boiling range 60-95°C
CAS: 64742-49-0 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.178 MDL Number: MFCD00081849 InChI Key: AFABGHUZZDYHJO-UHFFFAOYSA-N Synonym: isohexane,pentane, 2-methyl,2-methylpentan,2-methyl pentane,dimethylpropylmethane,1,1-dimethylbutane,iso-hexane,methyl pentane,unii-49ib0u6mld,2-methyl-pentane PubChem CID: 7892 SMILES: CCCC(C)C
Petroleum ether, ACS reagent, boiling range 35-60°C
CAS: 64742-49-0 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.178 MDL Number: MFCD00081849 InChI Key: AFABGHUZZDYHJO-UHFFFAOYSA-N Synonym: isohexane,pentane, 2-methyl,2-methylpentan,2-methyl pentane,dimethylpropylmethane,1,1-dimethylbutane,iso-hexane,methyl pentane,unii-49ib0u6mld,2-methyl-pentane PubChem CID: 7892 IUPAC Name: Petroleum ether SMILES: CCCC(C)C
Petroleum ether, ACS reagent, boiling range 40-60°C
CAS: 64742-49-0 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.178 MDL Number: MFCD00081849 InChI Key: AFABGHUZZDYHJO-UHFFFAOYSA-N Synonym: isohexane,pentane, 2-methyl,2-methylpentan,2-methyl pentane,dimethylpropylmethane,1,1-dimethylbutane,iso-hexane,methyl pentane,unii-49ib0u6mld,2-methyl-pentane PubChem CID: 7892 SMILES: CCCC(C)C
Petroleum ether, extra pure, boiling range 60-95°C
CAS: 64742-49-0 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.178 MDL Number: MFCD00081849 InChI Key: AFABGHUZZDYHJO-UHFFFAOYSA-N Synonym: isohexane,pentane, 2-methyl,2-methylpentan,2-methyl pentane,dimethylpropylmethane,1,1-dimethylbutane,iso-hexane,methyl pentane,unii-49ib0u6mld,2-methyl-pentane PubChem CID: 7892 SMILES: CCCC(C)C
Petroleum ether, Extra Dry, boiling range 40-60°C, water <50ppm, AcroSeal™
CAS: 64742-49-0 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.178 MDL Number: MFCD00081849 InChI Key: AFABGHUZZDYHJO-UHFFFAOYSA-N Synonym: isohexane,pentane, 2-methyl,2-methylpentan,2-methyl pentane,dimethylpropylmethane,1,1-dimethylbutane,iso-hexane,methyl pentane,unii-49ib0u6mld,2-methyl-pentane PubChem CID: 7892 IUPAC Name: Petroleum ether SMILES: CCCC(C)C
Petroleum Ether, BP, MilliporeSigma™
CAS: 64742-49-0
Petroleum ether, extra pure, boiling range 35-60°C
CAS: 64742-49-0 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.178 MDL Number: MFCD00081849 InChI Key: AFABGHUZZDYHJO-UHFFFAOYSA-N Synonym: isohexane,pentane, 2-methyl,2-methylpentan,2-methyl pentane,dimethylpropylmethane,1,1-dimethylbutane,iso-hexane,methyl pentane,unii-49ib0u6mld,2-methyl-pentane PubChem CID: 7892 IUPAC Name: Petroleum ether SMILES: CCCC(C)C
Petroleum ether, extra pure, boiling range 40-60°C, Thermo Scientific Chemicals
CAS: 64742-49-0 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.178 MDL Number: MFCD00081849 InChI Key: AFABGHUZZDYHJO-UHFFFAOYSA-N Synonym: isohexane,pentane, 2-methyl,2-methylpentan,2-methyl pentane,dimethylpropylmethane,1,1-dimethylbutane,iso-hexane,methyl pentane,unii-49ib0u6mld,2-methyl-pentane PubChem CID: 7892 SMILES: CCCC(C)C
Petroleum ether, for residue analysis, boiling range 40-60°C, ECD tested halocarbons free
CAS: 64742-49-0 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.178 MDL Number: MFCD00081849 InChI Key: AFABGHUZZDYHJO-UHFFFAOYSA-N Synonym: isohexane,pentane, 2-methyl,2-methylpentan,2-methyl pentane,dimethylpropylmethane,1,1-dimethylbutane,iso-hexane,methyl pentane,unii-49ib0u6mld,2-methyl-pentane PubChem CID: 7892 SMILES: CCCC(C)C
Petroleum Ether Non UV, MilliporeSigma™
CAS: 64742-49-0 MDL Number: MFCD00081849 IUPAC Name: Petroleum ether
Petroleum ether, for residue analysis, 40-60°C,ECD tested lindane/DDT<5ng/l, res <0.0003%
CAS: 64742-49-0 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.178 MDL Number: MFCD00081849 InChI Key: AFABGHUZZDYHJO-UHFFFAOYSA-N Synonym: isohexane,pentane, 2-methyl,2-methylpentan,2-methyl pentane,dimethylpropylmethane,1,1-dimethylbutane,iso-hexane,methyl pentane,unii-49ib0u6mld,2-methyl-pentane PubChem CID: 7892 SMILES: CCCC(C)C
Petroleum ether, for analysis, boiling range 80-110°C
CAS: 64742-49-0 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.178 MDL Number: MFCD00081849 InChI Key: AFABGHUZZDYHJO-UHFFFAOYSA-N Synonym: isohexane,pentane, 2-methyl,2-methylpentan,2-methyl pentane,dimethylpropylmethane,1,1-dimethylbutane,iso-hexane,methyl pentane,unii-49ib0u6mld,2-methyl-pentane PubChem CID: 7892 SMILES: CCCC(C)C
Petroleum ether, for residue analysis, 40-60°C, for trace anal. of polyarom. hydrocarbons
CAS: 64742-49-0 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.178 MDL Number: MFCD00081849 InChI Key: AFABGHUZZDYHJO-UHFFFAOYSA-N Synonym: isohexane,pentane, 2-methyl,2-methylpentan,2-methyl pentane,dimethylpropylmethane,1,1-dimethylbutane,iso-hexane,methyl pentane,unii-49ib0u6mld,2-methyl-pentane PubChem CID: 7892 IUPAC Name: Petroleum ether SMILES: CCCC(C)C
- 1
- 2