Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Petroleum ether, Laboratory Reagent, bp 40-60 °C, Solstice

Catalog No. 60090421 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
5 L
4 x 5 L
Packaging:
Aluminum bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-090-421 5 L Aluminum bottle
60-044-922 4 x 5 L Aluminum bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-090-421 Supplier Solstice Supplier No. 159675L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Laboratory Reagent, bp 40-60 °C

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64742-49-0
Molecular Formula C6H14
Molecular Weight (g/mol) 86.178
MDL Number MFCD00081849
InChI Key AFABGHUZZDYHJO-UHFFFAOYSA-N
PubChem CID 7892
IUPAC Name 2-methylpentane
SMILES CCCC(C)C

Specifications

Color Colorless
Density 0.65g/cu.cm at 20°C
Boiling Point 40°C to 60°C (1013 hPa)
Flash Point −30°C (−22°F)
Odor Characteristic
Packaging Aluminum bottle
Quantity 5 L
UN Number UN1268
Synonym isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane
Solubility Information Practically insoluble in water
Grade Laboratory Reagent
Physical Form Liquid
Chemical Name or Material Petroleum Ether
Show More Show Less
Hazard Statement P210-P301 + P310-P331-P370 + P378-P403 + P235
Precautionary Statement H225-H304-H315-H336-H411
DOTInformation class 3 - PG 2 - EHS - VP > 110kPa - Petroleum distillates, n.o.s., HI: S
EINECSNumber 265-151-9
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.