accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNOOHOHO

CAS RN 64769-66-0

IUPAC Name: pyrrolidine-2,4-dicarboxylic acid
Synonyms: pyrrolidine-2,4-dicarboxylic acid
Molecular Weight (g/mol): 159.14
Molecular Formula: C6H9NO4
InChi Key: NRSBQSJHFYZIPH-UHFFFAOYNA-N
SMILES: OC(=O)C1CNC(C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)