accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOSOOFFF

CAS RN 65007-00-3

IUPAC Name: pyridin-2-yl trifluoromethanesulfonate
Synonyms: pyridin-2-yl trifluoromethanesulfonate
Molecular Weight (g/mol): 227.16
Molecular Formula: C6H4F3NO3S
InChi Key: COLRMVLTWJTLFJ-UHFFFAOYSA-N
SMILES: FC(F)(F)S(=O)(=O)OC1=CC=CC=N1
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)