Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Pyridyl Trifluoromethanesulfonate 98.0+%, TCI America™
SDP

Catalog No. P18405G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P18405G Supplier TCI America Supplier No. P18405G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Organic chemical reagent used for synthetic research and chemical analysis.

Chemical Identifiers

CAS 65007-00-3
Molecular Formula C6H4F3NO3S
Molecular Weight (g/mol) 227.157
MDL Number MFCD00192531
InChI Key COLRMVLTWJTLFJ-UHFFFAOYSA-N
Synonym 2-pyridyl trifluoromethanesulfonate, 2-pyridyl triflate, pyridyl triflate, acmc-1bdfn, 2-pyridyltrifluoromethanesulfonate, 2-pyridyl trifluoromethane-sulfonate, pyridin-2-yltrifluoromethanesulfonate, 2-trifluoro-methylsulfonyloxy-pyridine, trifluoromethanesulfonic acid 2-pyridyl ester
PubChem CID 3947626
IUPAC Name pyridin-2-yl trifluoromethanesulfonate
SMILES C1=CC=NC(=C1)OS(=O)(=O)C(F)(F)F

Specifications

Color Yellow
Boiling Point 108°C
Quantity 5 g
UN Number 1993
Formula Weight 227.16
Percent Purity ≥98.0% (GC)
Grade Not Available
Physical Form Liquid
Chemical Name or Material 2-Pyridyl Trifluoromethanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.