accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6501-72-0

IUPAC Name: 2-(3-phenyl-4,5-dihydro-1,2-oxazol-5-yl)acetic acid
Synonyms: (3-phenyl-4,5-dihydro-1,2-oxazol-5-yl)acetic acid
Molecular Weight (g/mol): 205.21
Molecular Formula: C11H11NO3
InChi Key: MUFJHYRCIHHATF-UHFFFAOYNA-N
SMILES: OC(=O)CC1CC(=NO1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results