accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONOO

CAS RN 65039-18-1

IUPAC Name: 1-(dodecyloxy)-4-nitrobenzene
Synonyms: 1-(dodecyloxy)-4-nitrobenzene
Molecular Weight (g/mol): 307.43
Molecular Formula: C18H29NO3
InChi Key: VZOAOBWVYOLSII-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCOC1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results