CAS RN 6515-58-8
IUPAC Name:
3-(bromomethyl)benzoate
Synonyms:
3-(bromomethyl)benzoate
Molecular Weight (g/mol):
214.04
Molecular Formula:
C8H6BrO2
InChi Key:
FTPAHNNMUYOHOB-UHFFFAOYSA-M
SMILES:
[O-]C(=O)C1=CC=CC(CBr)=C1
1
–
1
of
1
results
1
Sigma Aldrich 3-(Bromomethyl)benzoic acid
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.